Learn More
Medchemexpress LLC [Des-Arg9]-Bradykinin acetate | 23827-91-0 | 99.9% | 964.07 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP0298A50MG
Encompass_Preferred
[Des-Arg9]-Bradykinin acetate is a Bradykinin B1 receptor agonist that displays selectivity for B1 over B2 receptors. The B2 receptor is responsible for the actions of bradykinin (BK) and lysyl-bradykinin (Lys-BK), which are formed in response to injury from kininogen precursors. The B1 receptor, however, mediates the actions of [Des-Arg9]-Bradykinin (des-Arg9-BK) and Lys-des-Arg Arg9-BK, which are formed by carboxypeptidases acting on BK and Lys-BK. This product is for research use only.
  • Bradykinin B1 receptor agonist
  • Displays selectivity for B1 over B2 receptors
  • Mediates actions of [Des-Arg9]-Bradykinin (des-Arg9-BK) and Lys-des-Arg Arg9-BK
  • For research use only
Catalog No. 50-004-25194