Learn More
Medchemexpress LLC Des-Gly10,D-Ala6,Pro-NHEt9-LHRH (Surfagon) | 52435-06-0 | 99.5% | 1167.32 | 1 MG

Supplier: Medchemexpress LLC HYP00481MG
(Des-Gly10,D-Ala6,Pro-NHEt9)-LHRH (Surfagon) is an agonist of gonadotropin-releasing hormone (GnRH). It serves as an internal standard for LC-MS analysis of leuprorelide acetate, with potential applications in biochemical analysis and fertility research.
- Agonist of gonadotropin-releasing hormone (GnRH)
- Used as an internal standard for LC-MS analysis of leuprorelide acetate
- Appearance: White to off-white solid
- Target: GnRH receptor
- Pathway: GPCR/G protein
- Storage (powder): -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.