Learn More
CPC Scientific H-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Glu-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GLUC005A
Encompass
ONE-LETTER SEQUENCE: SQGTFTSEYSKYLDSRRAQDFVQWLMNTMOLECULAR FORMULA: C148H220N40O48S1MOLECULAR WEIGHT:3358.68STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [110084-95-2], SYNONYMS: des-His1-[Glu9]-Glucagon (1-29) amideRESEARCH AREA: Diabetes
Catalog No. 50-210-9367