Learn More
CPC Scientific H-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Glu-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GLUC006B
Encompass
ONE-LETTER SEQUENCE: SQGTFTSEYSKYLDSRRAQDFVQWLMNT-NH2MOLECULAR FORMULA: C148H221N41O47S1MOLECULAR WEIGHT:3358.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [110084-95-2], RESEARCH AREA: DiabetesREFERENCES: C.G. Unson et al., Peptides, 10, 1171 (1989)
Catalog No. 50-210-9370