Learn More
Medchemexpress LLC Des(methylpiperazinyl-N-methyl) imatinib dimer impurity | 1365802-18-1 | MFCD28899574 | 96.1% | C52H48N12O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13127510MG
Encompass_Preferred
Imatinib Impurity E is an analytical reference standard used to characterize and quantify a known impurity of imatinib (CAS 1365802-18-1). Supplied as a high-purity compound (96.05%), it is intended for research and quality-control workflows such as impurity profiling, method development, and stability testing.
  • Serves as an analytical reference standard for impurity identification.
  • Supports HPLC, LC-MS, and method validation workflows.
  • Supplied with a certificate of analysis for traceability.
  • High reported purity suitable for quantitative analysis.
  • Available in small laboratory pack sizes for analytical use.
Catalog No. 50-002-61826