Learn More
Carbosynth LLC. Des-n-Octanoyl-[Ser3]-Ghrelin (Rat), 0.5mg, MW: (M.W. 3188.67), Molecular formula: C139H231N45O41
SDP
Supplier:  Carbosynth LLC. PGH3654PI0.5MG
Encompass_Preferred
Synonyms: H-Gly-Ser-Ser-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OHNon-Acylated Ghrelin (Rat), Application: Counteracts Metabolic Effects of Acylated Ghrelin, Storage Temperature: -20°C, References: H Hosoda, M Kojima, H Matsuo, and K Kangawa, Biochem. Biophys. Res. Commun., 279, 909 (2000)., F. Broglio, C. Gottero, F. Prodam, C. Gauna, G. Muccioli, M. Papotti, T. Abribat, A.J. Van der Lely, and E. Ghigo, J. Clin. Endoc. and Metabolism, 89, 3062 (2004)., C. Gauna, F.M. Meyler, J.A.M.J.L. Janssen, R.J.D. Delhanty, T. Abribat, P. van Koetsveld, L.J. Hofland, F. Broglio, E.Ghigo, and A.J. van der Lely, J. Clin. Endoc. and Metabolism, 89, 5035 (2004)
Catalog No. 50-168-6511