Learn More
Cayman Chemical 3 4 7TrIhydroxyIsoflavon 100mg
SDP
Supplier:  Cayman Chemical 20930100 MG
Encompass_Preferred
A cell-permeable copper-containing 3-formylchromone derivative that inhibits Akt in an array of cancer cells (IC50s 10-34 UM) also causes NF-KB inactivation in an orthotopic pancreatic tumor model using COLO 357 cells
Catalog No. 50-298-4170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.