Learn More
Cayman Chemical Desacetylcinobufagin 10MG
SDP
Supplier:  Cayman Chemical 3982210 MG
Encompass_Preferred
An active metabolite of cinobufagin; cytotoxic to HepG2, K562, and HL-60 cells (IC50s = 51.2, 50.1, and 7.5 µM, respectively)
Catalog No. 50-294-2375