Learn More
Bachem Neuropeptide Y (free acid) (human, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4031097.1000
Encompass_Preferred
Neuropeptide Y (free acid) (human, rat) Trifluoroacetate, H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-OH, 99575-89-0, C189H284N54O58S, Mr 4272.73, 1mg. NPY (free acid) (human, rat). Desamidation of neuropeptide Y suppresses almost all of NPY effects.
Catalog No. 50-194-6962