Learn More
Cayman Chemical desbutyl LumefntrIn 10mg
SDP
Supplier:  Cayman Chemical 2493510 MG
Encompass_Preferred
An active metabolite of lumefantrine; active against chloroquine-sensitive and -resistant strains of P. falciparum (IC50s = 9 and 9.5 nM, respectively); inhibits the hERG potassium channel in HEK293 cells expressing the human channel (IC50 = 5.49 µM)
Catalog No. 50-293-2991