Learn More
Medchemexpress LLC Deschloroetizolam | 40054-73-7 | 99.3% | C17H16N4S | 10MG

Supplier: Medchemexpress LLC HY13799410MG
Deschloroetizolam is a designer benzodiazepine research reference compound supplied as a high-purity analytical standard for laboratory and in vitro research. It is a psychoactive drug derivative intended for analytical and pharmacological studies and is not for human consumption. Safety documentation and a product datasheet are available.
- High purity: ~99.3% (product page and datasheet report 99.29% and 99.82%)
- Chemical identity: CAS 40054-73-7, formula C17H16N4S
- Package: supplied as a 10 mg vial for laboratory use
- Documentation: datasheet and SDS available in multiple languages
- Intended use: analytical method development and in vitro research
- Handling: for laboratory use only, follow safety and regulatory guidance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.