Learn More
Medchemexpress LLC Desciclovir | 84408-37-7 | 99.9% | 209.21 g·mol⁻¹ | C8H11N5O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10650625MG
Encompass_Preferred
Desciclovir is a potent, orally active prodrug of the antiviral acyclovir that is converted in vivo by xanthine oxidase. Supplied as an off-white to light-yellow solid for research use, it offers defined solubility and storage characteristics for reliable laboratory handling.
  • Potent, orally active prodrug of acyclovir converted in vivo by xanthine oxidase.
  • Off-white to light-yellow solid suitable for research use.
  • High purity (99.94%).
  • Soluble in DMSO (83.33 mg/mL) and water (25 mg/mL) with warming and sonication.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (2 years).
  • Available in small milligram pack sizes for laboratory experiments.
Catalog No. 50-002-40297