Learn More
Medchemexpress LLC Desciclovir | 84408-37-7 | MFCD00866932 | 99.9% | 209.21 | C8H11N5O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY106506100MG
Encompass_Preferred
Desciclovir is an orally active prodrug of acyclovir that is converted in vivo by xanthine oxidase, producing antiviral activity against herpesviruses. It is supplied as a high-purity research chemical suitable for in vitro and in vivo antiviral studies.
  • Converted to acyclovir in vivo by xanthine oxidase.
  • Suitable for antiviral research against herpesviruses.
  • High reported purity for consistent experimental results.
  • Available in multiple small-scale pack sizes for research workflows.
  • Molecular weight 209.21, formula C8H11N5O2.
Catalog No. 50-002-42092