Learn More
Medchemexpress LLC Desciclovir | 84408-37-7 | MFCD00866932 | 99.9% | 209.21 g/mol | C8H11N5O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10650610MG
Encompass_Preferred
Desciclovir is an orally active prodrug of the antiherpetic agent acyclovir used as a research reagent. It is converted in vivo to acyclovir by xanthine oxidase and has demonstrated activity against herpesviruses. The material is supplied as a powder and should be handled and stored per the safety data sheet recommendations.
  • Orally active prodrug of acyclovir.
  • Converted in vivo to acyclovir by xanthine oxidase.
  • High reported purity (approximately 99.9%).
  • Supplied as a powder suitable for laboratory formulation.
  • Recommended storage prolongs stability under frozen conditions.
Catalog No. 50-002-40295