Learn More
Medchemexpress LLC Desglymidodrine 10mg | 3600-87-1 | 10 MG

Supplier: Medchemexpress LLC HY11479410MG
Desglymidodrine is the active metabolite of midodrine and a selective α1-adrenoceptor agonist supplied as an off-white to light yellow solid for research use. It has molecular formula C10H15NO3, molecular weight 197.23 g/mol, and reported purity of 97.33%.
- Selective α1-adrenoceptor agonist.
- Active metabolite of midodrine.
- Off-white to light yellow solid form.
- Molecular formula C10H15NO3 and molecular weight 197.23 g/mol.
- Purity reported as 97.33%.
- Store at 4°C and protect from light; in solution store frozen as recommended.
- Supplied as a 10 mg research reagent suitable for in vitro studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.