Learn More
Electron Microscopy Sciences Desiccator Plate 230 mm
SDP
Supplier:  Electron Microscopy Sciences 7124223
Encompass_Preferred
These 4.8 mm thick plates are able to withstand temperatures up to 150 degree C (300 degree F) and have excellent resistance to thermal shock, and to chemicals. They are non-combustible and asbestos free. Round plates are perforated with 4 mm(.165") diameter holes on 12.7 mm (1/2") centers in a straight line pattern.
Catalog No. 50-189-9542