Learn More
Medchemexpress LLC Desipramine hydrochloride | 58-28-6 | 99.76% | 50 MG

Supplier: Medchemexpress LLC HYB127250MG
Desipramine hydrochloride is a potent inhibitor of norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT). It is primarily used in research due to its specific inhibitory activities, with reported Ki values of 4, 61, and 78,720 nM for NET, SERT, and DAT respectively. This high-purity compound (99.76%) is supplied for various in vitro and in vivo applications, and its properties and uses are well-documented in scientific literature.
- Inhibits norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT).
- High purity of 99.76%.
- Molecular weight of 302.84.
- Chemical formula is C18H23ClN2.
- Appears as a solid, white to off-white in color.
- Detailed solubility information is available for diverse experimental setups.
- Frequently cited in scientific publications for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.