Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desipramine hydrochloride | 58-28-6 | 99.8% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYB127210MM/1ML

Encompass_Preferred

Desipramine hydrochloride is a chemical compound primarily known for its inhibitory effects on norepinephrine, 5-HT, and dopamine transporters. It is provided as a solution, ensuring high purity for research applications.

  • Inhibits norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT).
  • Has a purity of 99.76%.
  • Molecular weight is 302.84.
  • Chemical formula is C18H23ClN2.
  • Appears as a white to off-white solid.
  • Available as a 10 mM solution in 1 mL of DMSO.
  • Requires sealed storage at 4°C, away from moisture. In solution, store at -80°C for 1 year or -20°C for 6 months.

Catalog No. 50-004-11563


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.