Learn More
Medchemexpress LLC Desipramine hydrochloride | 58-28-6 | MFCD00058108 | 1 G
Supplier: Medchemexpress LLC HYB12721G
Desipramine hydrochloride acts as an inhibitor for norepinephrine transporter (NET), 5-HT transporter (SERT), and dopamine transporter (DAT). It exhibits Ki values of 4, 61, and 78,720 nM for NET, SERT, and DAT respectively. Studies have shown that chronic administration can reduce NET expression in a dose-dependent manner.
- Inhibitor of norepinephrine transporter (NET)
- Inhibitor of 5-HT transporter (SERT)
- Inhibitor of dopamine transporter (DAT)
- Reduces norepinephrine transporter expression in a dose-dependent manner