Learn More
Medchemexpress LLC Desisopropyl atrazine-d5 | 1189961-78-1 | 98.0% | 178.63 | C5H3D5ClN5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146604S5MG
Encompass_Preferred
Desisopropylatrazine-d5 is the deuterium-labeled isotopologue of desisopropylatrazine (CAS 1189961-78-1), provided as an analytical reference standard for research and isotope-dilution mass spectrometry. Molecular formula C5H3D5ClN5; molecular weight 178.63 g·mol⁻¹. Chemical purity is typically reported around 98%.
  • Isotopically labeled (d5) analog for use as an internal standard.
  • Suitable for isotope-dilution quantitation by GC-MS or LC-MS.
  • Chemical purity approximately 98% for analytical use.
  • Molecular formula C5H3D5ClN5 and molecular weight 178.63 g·mol⁻¹.
  • Supplied as a neat solid in small research quantities.
  • For research use only; not intended for human or clinical use.
Catalog No. 50-002-22643