Learn More
Medchemexpress LLC Desloratadine | 100643-71-8 | 100.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB053910G
Encompass_Preferred
Desloratadine is an orally active and selective H1 receptor antagonist (Ki=0.9 nM) with anti-inflammatory and anti-allergic activities. It inhibits the release of histamine and LTC4 from human basophils, modulating regulatory signals for IL-4 and IL-13 production. This compound effectively alleviates seasonal allergic rhinitis (SAR) symptoms in patients with concurrent asthma, making it valuable for research into allergic rhinitis and chronic idiopathic urticaria.
  • Orally active and selective H1 receptor antagonist
  • Exhibits anti-inflammatory and anti-allergic properties
  • Inhibits histamine and LTC4 release from basophils
  • Modulates IL-4 and IL-13 production
  • Alleviates seasonal allergic rhinitis symptoms
  • Used in studies for allergic rhinitis and chronic idiopathic urticaria
  • Appears as an off-white to light yellow solid
  • Molecular weight: 310.83
Catalog No. 50-004-05486