Learn More
TARGETMOL CHEMICALS INC Desloratadine 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2520200MG
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Desloratadine (Sch34117) is a long-acting piperidine derivate with selective H1 antihistaminergic and non-sedating properties. Desloratadine diminishes the typical histaminergic effects on H1-receptors in bronchial smooth muscle, capillaries and gastrointestinal smooth muscle, including vasodilation, bronchoconstriction, increased vascular permeability, pain, itching and spasmodic contractions of gastrointestinal smooth muscle. Desloratadine is used to provide symptomatic relieve of allergic symptoms. Purity 100%
Catalog No. 50-287-5882