Learn More
Cayman Chemical DesloratdIn-d4 500ug
SDP
Supplier:  Cayman Chemical 26776500 UG
Encompass_Preferred
An internal standard for the quantification of desloratadine by GC- or LC-MS
Catalog No. 50-293-2954