Learn More
Cayman Chemical DesloratdIn-d4 5mg
SDP
Supplier:  Cayman Chemical 267765 MG
Encompass_Preferred
An internal standard for the quantification of desloratadine by GC- or LC-MS
Catalog No. 50-293-5167