Learn More
Cayman Chemical DesmethoxyyngonIn 5mg
SDP
Supplier:  Cayman Chemical 238145 MG
Encompass_Preferred
An analytical reference standard categorized as a kavalactone; increases the duration of sleep in a mouse model of pentobarbital-induced sleep; intended for research and forensic applications
Catalog No. 50-299-4852