Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desmethyl celecoxib | 170569-87-6 | MFCD03453540 | 99.4% | 367.35 | C16H12F3N3O2S | 1 ML
SDP

Supplier:  Medchemexpress LLC HY11813910MM 1ML

Encompass_Preferred

Desmethyl celecoxib is a selective cyclooxygenase-2 (COX-2) inhibitor and an analog of celecoxib, used as a research reagent for COX-2 modulation and inflammation studies. Offered as a 10 mM solution in DMSO (1 mL) or as solid powder formats.

  • Selective COX-2 inhibitor (IC50 = 32 nM).
  • Supplied as 10 mM solution in DMSO, 1 mL.
  • High reported purity (99.42%).
  • Molecular weight 367.35 g/mol.
  • Appearance: white to off-white solid; solution ready to use.
  • Recommended storage: powder -20°C for long term; in solvent -80°C for best stability.

Catalog No. 50-003-07836


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.