Learn More
Cayman Chemical DesmethylclotIazepam 1mg
SDP
Supplier:  Cayman Chemical 278381 MG
Encompass_Preferred
An analytical reference standard categorized as a thienodiazepine; an active metabolite of clotiazepam; intended for research and forensic applications
Catalog No. 50-294-0273