Learn More
Cayman Chemical Desmethylen ParoxetInhydr 10mg
SDP
Supplier:  Cayman Chemical 1590810 MG
Encompass_Preferred
A major urinary metabolite of the selective serotonin reuptake inhibitor paroxetine; may be used as a control in LC/MS or GC/MS paroxetine testing applications
Catalog No. 50-296-9770