Learn More
Genetex Inc Desmin Monoclonal Antibody [MSVA-651M] HistoMAX™ (GTX04410) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04410
Encompass_Preferred
The Desmin antibody [MSVA-651M] HistoMAX™ (GTX04410) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human rhabdomyosarcoma (RMS) tissue using GTX04410 Desmin antibody [MSVA-651M] HistoMAX. Strong desmin positivity of all tumor cells of a rhabdomyosarcoma.
Catalog No. NC3666783