Learn More
ABclonal Technology Desmin Rabbit pAb
SDP
Supplier:  ABclonal Technology A1116500UL
Encompass_Preferred
This gene encodes a muscle-specific class III intermediate filament. Homopolymers of this protein form a stable intracytoplasmic filamentous network connecting myofibrils to each other and to the plasma membrane. Mutations in this gene are associated with desmin-related myopathy, a familial cardiac and skeletal myopathy (CSM), and with distal myopathies.
Catalog No. 50-001-31006