Learn More
ABclonal Technology Desmocollin 1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A23580100UL
Encompass_Preferred
The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily These desmosomal family members along with the desmogleins are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation A subtype of IgA pemphigus a life-threatening autoimmune disease is characterized by the presence of autoantibodies that target the encoded protein The desmosomal family members are arranged in two clusters on chromosome 18 Alternative splicing of this gene results in multiple transcript variants At least one of these variants encodes a preproprotein that is proteolytically processed to generate the mature protein
Catalog No. 50-000-06233