Learn More
Genetex Inc Desmoglein 1 Monoclonal Antibody [MSVA-544M] HistoMAX™ (GTX04464) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04464
Encompass_Preferred
The Desmoglein 1 antibody [MSVA-544M] HistoMAX™ (GTX04464) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skin tissue using GTX04464 Desmoglein 1 antibody [MSVA-544M] HistoMAX. Squamous epithelium of the skin showing a strong membranous desmoglein 1 immunostaining.
Catalog No. NC3667747