Learn More
Genetex Inc Desmoglein 3 Monoclonal Antibody [MSVA-543M] HistoMAX™ (GTX04468) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04468
Encompass_Preferred
The Desmoglein 3 antibody [MSVA-543M] HistoMAX™ (GTX04468) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human tonsil tissue using GTX04468 Desmoglein 3 antibody [MSVA-543M] HistoMAX. In the tonsil a moderate to strong desmoglein 3 immunostaining occurs in squamous epithelial cells of crypts. All other cell types are desmoglein 3 negative.
Catalog No. NC3667757