Learn More
Cayman Chemical Desogestrel-13C2-d2 1mg
SDP
Supplier:  Cayman Chemical 307641 MG
Encompass_Preferred
An internal standard for the quantification of desogestrel by GC- or LC-MS
Catalog No. 50-298-8975