Learn More
Apexbio Technology LLC Desogestrel 54024-22-5 25mg
SDP
Supplier:  Apexbio Technology LLC B497625
Encompass_Preferred
Desogestrel (CAS 54024-22-5) is a synthetic progestogen derivative targeting the progesterone receptor It is designed to selectively activate this receptor thereby suppressing gonadotropin secretion and inhibiting ovarian follicle maturation and ovulation Desogestrel exerts its biological activity primarily through selective activation of progesterone receptors Based on its chemical structure desogestrel contains an alkyne functional group that enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions with azide-containing molecules Based on these pharmacological properties desogestrel holds research potential in targeted drug modification probe labeling and pharmacological investigations as well as serving as an active ingredient in oral contraceptive formulations
Catalog No. 50-000-91872