Learn More
Medchemexpress LLC Desogestrel | 54024-22-5 | MFCD00869346 | 310.47 | C22H30O | 50MG

Supplier: Medchemexpress LLC HY1251650MG
Desogestrel is a third-generation progestin supplied as an analytical reference standard for use in qualitative and quantitative method development. It is provided as a powder formulation intended for use in HPLC, GC, and MS assays; a Certificate of Analysis documents purity and recommended storage conditions.
- Analytical reference standard suitable for HPLC, GC, and MS.
- CAS number 54024-22-5.
- Molecular formula C22H30O; molecular weight 310.47.
- Powder appearance for laboratory use.
- Available in small package sizes for analytical testing, including 50 mg.
- Certificate of Analysis available for purity and storage information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.