Learn More
Apexbio Technology LLC Desonide 638-94-8 50mg
SDP
Supplier:  Apexbio Technology LLC B215850
Encompass_Preferred
Desonide (CAS 638-94-8) is a nonfluorinated topical corticosteroid targeting intracellular corticosteroid (glucocorticoid) receptors It is designed to modulate inflammatory pathways thereby regulating cytokine expression inflammatory cell migration and dermal vascular tone Desonide exerts its biological activity primarily through receptor-mediated transcriptional changes that decrease the synthesis and release of pro-inflammatory mediators suppress inflammatory cell migration and induce vasoconstriction Based on these pharmacological properties Desonide holds research potential in experimental models of dermatological inflammation dermatitis and cutaneous immune responses especially in studies exploring corticosteroid-mediated signaling and dermal pharmacokinetics/dynamics
Catalog No. 50-000-91007