Learn More
Medchemexpress LLC Desonide | 638-94-8 | 99.2% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0248500MG
Encompass_Preferred
Desonide is a non-fluorinated corticosteroid anti-inflammatory agent that acts on the glucocorticoid receptor. It can also specifically bind to the mutant huntingtin protein (mHTT), reducing its level and toxicity. Desonide is used in the research of Huntington's disease and inflammatory diseases such as atopic dermatitis.
  • Non-fluorinated corticosteroid anti-inflammatory agent
  • Acts on the glucocorticoid receptor
  • Binds to mutant huntingtin protein (mHTT)
  • Reduces mHTT levels and toxicity
  • Used in research for Huntington's disease
  • Used in research for inflammatory conditions such as atopic dermatitis
Catalog No. 50-004-05589