Learn More
Medchemexpress LLC Desthiobiotin-NHS ester | 80750-24-9 | MFCD08062086 | 98.0% | 311.33 g/mol | C14H21N3O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW437745100MG
Encompass_Preferred
Desthiobiotin-NHS ester is a desthiobiotin analog bearing an N-hydroxysuccinimide (NHS) ester that selectively reacts with primary amines to form stable amide bonds. The desthiobiotin moiety binds streptavidin with lower affinity than biotin, allowing reversible affinity capture and mild elution, making it useful for labeling, enrichment, and probe development in biochemical assays.
  • Selective labeling of primary amines.
  • Enables reversible streptavidin-based affinity capture.
  • Forms stable amide bonds for durable conjugation.
  • Suitable for protein labeling and pull-down assays.
  • Available in multiple package sizes for research scalability.
Catalog No. 50-002-81145