Learn More
Brady™ Destructible Tamper-evident Matte Vinyl Labels
Identify product tampering and prevent one-piece label removal with the B-351 tamper-resistant vinyl label material. Easily print almost anywhere with a Brady™ portable label printer.
Supplier:  Brady™ M520351
 View more versions of this product
Catalog No. 17-012-256
Add to Cart
Description
Description
  • For use in asset tracking, rating plates, product security and tamper identification
  • Designed to fracture to show signs of product tampering and prevent one-piece removal
  • Made with a permanent adhesive that bonds tightly to almost any surface
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White
Die-cut
Top
Label
Asset tracking;Security identification
1
M5-20-351
160
-40°F to 176°F
0.0028 in
25.40 mm (W) x 50.80 mm (H)
1 in
25.40 mm
0.5 in.
M5-20-351
2011/65/EU, 2015/863
None
Matte
Vinyl
Thermal Transfer
Non-sterile
BMP51;BMP53;M511;M510
-40°C
160 Label(s) / Box
25.40 mm (W) x 50.80 mm (H)
-40°C to 80°C
0.071 mm
2 in.
25.40 mm
2 in
50.80 mm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov