Learn More
ATCC Desulfovibrio desulfuricans; subsp. desulfuricans
Supplier:  ATCC 27774
Encompass
Desulfovibrio desulfuricans subsp. desulfuricans strain MB is a whole-genome sequenced bacterium that was isolated from the rumen of sheep. This product produce restriction endonuclease DdsI and iron-sulfur protein and is capable of utilizing nitrate as well as sulfate during the anaerobic fermentation of lactate.
Catalog No. 50-238-0194