Learn More
Cayman Chemical Desvaleryl Valsartn 10mg
SDP
Supplier:  Cayman Chemical 3563410 MG
Encompass_Preferred
A potential impurity found in commercial preparations of valsartan; a degradation product formed under acidic conditions
Catalog No. 50-290-9901