Learn More
Cayman Chemical DETA NONOate, 146724-94-9, 100 mg
SDP
Supplier:  Cayman Chemical 82120100
Encompass_Preferred
Molecular Formula C4H13N5O2, Purity ≥98%, Formula Weight 163.2, Synonyms: (Z)-1-[N-(2-aminoethyl)-N-(2-ammonioethyl)amino]diazen-1-ium-1,2-diolate, Diethylenetriamine NONOate, NOC-18; DETA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 20 hours and 56 hours at 37C and 22-25C, pH 7.4, respectively, to liberate 2 moles of NO per mole of parent compound.{1190,5437}
Catalog No. NC9382295