Learn More
Cayman Chemical DETA NONOate, 146724-94-9, 500 mg
SDP
Supplier:  Cayman Chemical 82120500
Encompass_Preferred
DETA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 20 hours and 56 hours at 37°C and 22-25°C, pH 7.4, respectively, to liberate 2 moles of NO per mole of parent compound.{1190,5437}
Catalog No. NC1414735