Learn More
HOTSTICK USA INC AC Hotstick Voltage Detector
Supplier:  HOTSTICK USA INC BN 9005/02
Encompass_Preferred
he AC HotStick is a safety device designed for professional rescuers which provides warning of exposed high voltage AC from a safe distance. The AC HotStick will give early audible ("beeping") and visual (flashing LED) warning of the presence of dangerous voltages without the need to contact the surface carrying the current. The closer the user comes to the source of the voltage, the more rapidly the unit beeps and flashes.
Catalog No. NC9603012