Learn More
Medchemexpress LLC Deterenol hydrochloride | 23239-36-3 | MFCD28898982 | 99.7% | 231.72 | C11H18ClNO2 | 10 MG

Supplier: Medchemexpress LLC HYW70202710MG
Deterenol hydrochloride is a selective beta-adrenergic agonist supplied as the hydrochloride salt for research use. It produces vasodepressor effects and may cause increased heart rate, hypotension, palpitations, anxiety, and respiratory distress; handle with appropriate laboratory safety precautions.
- Selective beta-adrenergic agonist for pharmacological research.
- Hydrochloride salt, solid form for stable handling.
- High purity suitable for research applications (99.7%).
- Molecular formula C11H18ClNO2 and molecular weight 231.72.
- CAS number 23239-36-3 for unambiguous identification.
- Recommended sealed storage at -20°C; in solvent -80°C for 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.