Learn More
Cayman Chemical DeterenolhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3958310 MG
Encompass_Preferred
A β-adrenergic receptor agonist; increases tension in isolated right and left guinea pig atria and trachea (IC50s = 0.411, 1.44, and 8.37 µM, respectively), which endogenously express β-ARs, but has no effect on isolated rabbit aortic strips, which express α-ARs; induces lipolysis in human adipocytes
Catalog No. 50-291-8410