Learn More
Essendant Tide Plus Bleach Alternative Liquid Laundry Detergent, HE Compatible, 16 Loads, Original Scent, 25 oz Bottle, 6/Carton
Supplier:  Essendant PGC37373
Encompass_Preferred
Iconic liquid detergent infused with a chlorine-free bleach alternative keeps whites bright and colors vivid Active enzymes effectively break down and remove stains Rinses clean with a pleasant scent HE Compatible High Efficiency Designed for HE machines Washes 16 Loads Application- HE Laundry Detergent Applicable Material- Fabric Chemical Compound- Sodium and MEA Laureth Sulfate Sodium and MEA C10-16 Alkylbenzenesulfonate C10-16 Pareth C10-16 Alkyldimethylamine Oxide Sodium Borate Sodium Cumenesulfonate Calcium Formate Sodium and MEA Salts of C12-18 Fatty Acids Sodium and MEA Citrate Subtilisin Amylase Enzyme Mannanase Enzyme Polyethyleneimines Alkoxylated Pentasodium Pentetate Sodium Formate Fluorescent Brightener 71 Diethylenetriamine Methyl Di-t-Butyl Hydroxyhydrocinnamate Propylene Glycol Alcohol Ethanolamine Colorants Fragrances Water Dirt Types- Laundry Soils
Catalog No. 50-000-29245