Learn More
LABSYSTEMS DIAGNOSTICS OY Determination of Vitamin A/E
Supplier:  LABSYSTEMS DIAGNOSTICS OY 7250033
Encompass_Preferred
Vitamin A/E kit is used in routine clinical laboratories for the quantitative determination ofvitamin A and E in serum and plasma to detect deficiencies or hypervitaminosis. The separation of the vitamins and their specific internal standards is performed on a specific analytical column and all the analytes are measured by UV detection.
Catalog No. NC2865537