Learn More
Medchemexpress LLC Detomidine hydrochloride | 90038-01-0

Supplier: Medchemexpress LLC HYB0163A100MG
Detomidine hydrochloride is an imidazole derivative and a potent α2-adrenergic agonist that produces dose-dependent analgesic effects. This product is for research use only and has not been fully validated for medical applications.
- Potent α2-adrenergic agonist
- Produces dose-dependent analgesic effects
- Imidazole derivative
- Targets adrenergic receptor
- Involves GPCR/G protein and neuronal signaling pathways
- Molecular formula: C12H15CIN2
- Molecular weight: 222.71
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.